Bel-Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer
Resistant to chemicals sink strainer has four strong magnets in each corner to attract and hold Spinbar magnetic stirring bars. Bel Art™ SP Scienceware™ Spinbar™ Magnetic Sink Strainer is a simple solution to prevent your Spinbar™ Magnetic Stirring Bars from going down the sink drain.
Supplier:  Bel-Art™ F377870001
 View more versions of this product
Catalog No. 14-513-201
Explore available promotions
Description
Description
  • Long slits in the sides and skirt allow fluids to drain easily and rapidly
  • A wide skirt enables strainer to be used in a variety of sink drains; flat edge on one side allows it to be placed close to the sink wall
  • Four legs on the underside anchor the strainer firmly in sink drain and prevent it from being dislodged by flowing liquid
  • Color choices improve visibility of stirring bars
Specifications
Specifications
4 strong magnets in each corner, long slits in the sides and wide skirts
Spinbar™ Magnetic Sink Strainer
Stirring bars
Prevents stir bar from washing down drain
Green