Bel-Art™ SP Scienceware™ Spray Pump Bottles
Low-density polyethylene plunger style bottle delivers a fine mist spray
Supplier:  Bel-Art™ F116330000
 View more versions of this product
Catalog No. 01-189-0100
Explore available promotions
Description
Description
Low-density polyethylene plunger style bottle delivers a fine mist spray
  • Includes clear cap to help prevent evaporation and accidental leaks
  • 12/Pk.
Specifications
Specifications
8 oz., 250 mL
250 mL
8 oz.
LDPE