missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Standard Stem Funnels

Feature an exclusive filter-step internal design for speedier filtrations

$243.12 - $243.12

Specifications

Material Polypropylene
Autoclavable Autoclavable
Length (Metric) Stem 6.5cm
View More Specs

Products 1
Catalog Number Mfr. No. Length (Metric) Stem Diameter (Metric) Filter Price Quantity  
Catalog Number Mfr. No. Length (Metric) Stem Diameter (Metric) Filter Price Quantity  
22-109769
View Documents
Bel-Art™
F146760055
6.5cm 9cm
Pack of 12 Each for $243.12
Case of 3 Pack for $729.36
 
Description

Description

  • External ribs prevent airlock
  • The parallel stem measures 8mm O.D.
  • Molded polypropylene is steam autoclavable at 121°C (250°F)

Specifications

Specifications

Polypropylene
Autoclavable
6.5cm
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'