Learn More
Bel-Art™ SP Scienceware™ Sterileware™ Sampling Spatulas, with 6 in. V-shaped channel, Nonsterile
Ideal for every type of powdered or granulated material. Bel Art™ SP Scienceware™ Sterileware™ Sampling Spatulas are perfect for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
Supplier: Bel-Art™ F378400000
Description
Ideal for every type of powdered or granulated material. Bel Art™ SP Scienceware™ Sterileware™ Sampling Spatulas are perfect for taking sterile samples during process quality inspections and for piercing plastic-lined paper bags or similar packages.
Time-honored V-shape makes transferring powdered, flaked, crystalline or granular media a breeze
Long, 14 in. spatula (16-241-210) is perfect for dislodging caked or crystallized reagents from deep within tall bottles or vessels having a neck diameter as small as 0.75 in. (18mm)
Sterileware (16-241-210, 14-385-20)
- 9 and 14 in. models
- High-impact polystyrene
- Individually packaged
- Sterilized by gamma irradiation
- Lot stamped, shipped with Certificate of Processing
- 9 in. spatula
- Polypropylene
Specifications
Non-sterile | |
Sterileware™ Sampling Spatula | |
Ideal for every type of powdered or granulated material, perfect for taking sterile samples during process quality inspections and for piercing Plastic-lined Paper bags or similar package | |
0.75 in. | |
9 x 0.75 in. (22.9 x 1.9 cm) | |
2.99 in. | |
Polystyrene |
Single-use | |
Containers, bags and drums | |
9 in. | |
1.9 cm | |
6 in. L V-shaped channel | |
7.6 cm | |
22.9 cm |