Bel-Art™ SP Scienceware™ Sterileware™ Scoop An' Bag™ Sterile Sampler
Sample collection system includes a sterile 60mL (2 oz.) sampling scoop and sterile 11.4 x 22.9cm (4.5 x 9 in. ), 540mL (18.3 oz.) volume polyethylene twirl-type bag with large label area to hold the sample
Supplier: Bel-Art™ H369100000
Description
- One-piece scoop made from white, high-impact, FDA grade polystyrene plastic
- Long handle keeps the hand away from the sample
- Flat bottom allows scoop to remain upright for use as a weighing boat or to be placed on a flat surface
- 50 individually sealed and sterilized wrapped sets per box ready for one time use and disposal sterile
- Lot stamped for accurate lot tracing; shipped with a Certificate of Processing for Sterilization
Order Info
Sampling system comprised of a 2 oz. (60mL) white polystyrene scoop and a 9L x 4 1/2 in.W (228.6 x 114.3mm) polyethylene bag with a twirl-type closure packaged together.
Specifications
Scoop An Bag™ Sampler | |
Sterile | |
Ideal for food, pharmaceutical and environmental sampling |
60mL white polystyrene flat bottom one-piece sampling scoop with 12.74cm long handle and 7.8 L x 4.1 W x 2.9cm H bowl, 540mL 11.4 x 22.9cm polyethylene twirl-type bag with large label area to hold the sample | |
FDA |