missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ Stirring Rod

The solid, polypropylene stirring rod features a rounded end

Supplier:  Bel-Art™ F377660008

Catalog No. 22-216-010



Explore available promotions

Description

Description

The solid, polypropylene stirring rod features a rounded end

  • Steam autoclavable at 121°C (250°F) to avoid contamination
  • Diameter is 0.189 in. (4.7mm)
  • Length is 8 in. (20.3cm)

Specifications

Specifications

Autoclavable
Stirring Rod
0.189 in.
Rounded end
20.3 cm
Blunt End
8 in.
0.47 cm
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions