Bel-Art™ Swing Jaw™ Tubing Clamps
Provide nonslip closures for flexible tubing
Supplier: Bel-Art™ H182210000
Description
- Made of zinc-plated steel
- Knurled adjusting nut bears on a swinging jaw which permits full opening without removal
- Removable from tubing without disconnecting apparatus
- Smooth bearing surfaces will not damage tubing; tubing cannot slip out of clamp
- Strong enough to use with heavy wall vacuum lines
Specifications
Tubing Clamp | |
15mm (1/2 in.) O.D flexible tubing | |
Swing |
Zinc-plated Steel | |
Silver |