missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ Touch Free™ Automatic Waste Cans
Automatically open and close without a touch, protecting users from waste receptacle contact.
Supplier: Bel-Art™ F132020050
Includes: Each unit is supplied with a brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications).
Requires : Requires four D batteries (not included) which will provide approximately 4000 openings. All cans should be lined with disposable trash or biohazard bags (sold separately). Batteries and bags available on www.fishersci.com.
Description
Automatically open and close without a touch, protecting users from waste receptacle contact.
- When a hand or solid object is moved over the top, waste can detects motion, opens lid to accept waste, and then closes automatically
- Fitted well helps to reduce odor and airborne contamination
- Durable, dependable construction accommodates Bel-Art™ liners, biohazard liners or commercially available trash bags
- ON-OFF switch deactivates AUTO-OPEN/CLOSE function
- 2 Gallon (Cat. No. 11-394-451)
- Ideal for benchtop use
- Removable rigid autoclavable polypropylene liner will not leak or rust
- Body and lid made from easy-to-clean, molded ABS plastic
- White pail with white top
- Line with 14 x 19 in. biohazard disposal bags or suitable size standard trash bags
- Pail made from polypropylene with an ABS plastic top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Gray pail with gray top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- 7.3 Gallon (Cat. No. 03-409-55)
- Pail made from polypropylene with an ABS plastic top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Red pail with red top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- 12 Gallon (Cat. No. 11-394-453)
- Small footprint enables a fit into tight spaces
- Stainless-steel pail with black ABS plastic top
- Line with 24 x 30 in., 24 x 36 in. or 25 x 35 in. biohazard disposal bags or suitable size standard trash bags
- Prior to use, cans should be appropriately labeled with one of the included labels to indicate the type of waste to be placed inside the can.
Specifications
50 L | |
Silver | |
67.3 cm | |
29.3 cm | |
Brightly colored biohazard label and three other labels (recyclables, trash, and a blank label for individual applications) | |
Automatic Waste Can |
13.2 gal. | |
42 x 29.3 x 67.3 cm (16.5 x 11.5 x 26.5 in.) | |
42 cm | |
Autoclavable | |
Stainless Steel |