Bel-Art™ SP Scienceware™ Universal Animal Restrainer
For restraining small animals and rodents
Supplier:  Bel-Art™ H464000001
 View more versions of this product
Catalog No. 10-987-046
Explore available promotions
Description
Description
  • Fabricated of rigid, clear acrylic plastic with removable head and tail gates
  • Head gate comes with breathing holes and fits into any of four positions for various sizes of rodents
  • Tail gate allows maximum closeness of occluding cuff to body
  • For large rats and small guinea pigs
  • Inner raised floor may be removed for larger rodents
  • Nylon thumb screws serve as base fasteners and legs
Specifications
Specifications
Mice
Acrylic
10 to 40 g
Universal Animal Restrainer
Small