Bel-Art™ SP Scienceware™ Universal Animal Restrainer
For restraining small animals and rodents
Supplier: Bel-Art™ H464000002
Description
- Fabricated of rigid, clear acrylic plastic with removable head and tail gates
- Head gate comes with breathing holes and fits into any of four positions for various sizes of rodents
- Tail gate allows maximum closeness of occluding cuff to body
- For large rats and small guinea pigs
- Inner raised floor may be removed for larger rodents
- Nylon thumb screws serve as base fasteners and legs
Specifications
Rats and Hamsters | |
Acrylic | |
150 to 300 g |
Universal Animal Restrainer | |
Medium |