Bel-Art™ SP Scienceware™ Vakuwash™ Spectrophotometer Cell Washer
Cuvet washing device cleans without damaging
Supplier:  Bel-Art™ F389600000
Catalog No. 14-385-937
Explore available promotions
Includes: 500mL polypropylene filtering flask, Vikem™ stopper and PVC tube to connect to vacuum source.
Description
Description
Cuvet washing device cleans without damaging
  • Simple to use cleaner connects to any vacuum source
  • Partially fill funnel with cleaning solution and press open end of cell (or tube or vial) against soft rubber seal to force cleaning solution to spray into cell
  • Used fluids collect in flask for reuse
  • Continued pressure against cell creates airflow for drying
  • Use acetone as final wash before drying to help remove all water droplets
  • Minimum cuvet, tube or vial O.D. must be 9mm to form seal over the spray nozzle
Specifications
Specifications
Plastic
500 mL polypropylene vacuum flask, Vikem™ vinyl stopper and PVC tube to connect to vacuum source
50 Hz
Glassware Washer
230 V