missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Benchtop Coolers
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.
Supplier: Thermo Scientific™ 355501
Description
Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.
- Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
- Holds 12, 20 or 32 tubes
- Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
- Printed grid on one-way keyed lid for proper orientation and sample identification
- Molded of impact-resistant polycarbonate with locking handle for long-term use
- Stackable to save space
Specifications
133 mm | |
226 mm | |
165 mm | |
8.94 x 6.5 x 8.25 in. (226 x 165 x 133 mm) | |
4 x 5 | |
Non-filled/clear lid | |
Benchtop Cooler | |
0.2 to 0.5 mL | |
-20°C |
8.25 in. | |
8.94 in. | |
6.5 in. | |
4x5 compartments, -20 deg. | |
<-15°C, 1 | |
Polycarbonate | |
Clear | |
20 Tubes (0.2 to 0.5 mL) |