missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.

Supplier:  Thermo Scientific™ 51150032

 View more versions of this product

Catalog No. 15-350-51

Explore available promotions

Description

Description

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications

Specifications

146 mm
243 mm
157 mm
9.56 x 6.19 x 5.75 in. (243 x 157 x 146 mm)
4 x 8
Gel-filled/white lid
Benchtop Cooler
0.2 to 0.5 mL
-20°C
5.75 in.
9.56 in.
6.19 in.
4x8 compartments, gel filled, -20 deg,
<-15°C, 2
Polycarbonate
Blue, White
32 Tubes (0.2 to 0.5 mL)
Videos
SDS
Documents

Documents

Promotions

Promotions