missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Benchtop Coolers

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using benchtop coolers.

Supplier:  Thermo Scientific™ DS51160012

 View more versions of this product

Catalog No. 15-350-53


Explore available promotions

Description

Description

Protect enzymes, cells, reagents and solutions by maintaining freezer temperatures while on the laboratory bench using Thermo Scientific™ Benchtop Coolers.

  • Maintain freezer temperature during frost-free cycles, power failures and accidental door openings
  • Holds 12, 20 or 32 tubes
  • Inserts are included for 1.5 mL tubes to have the compatibility to hold 0.2-0.5 mL microcentrifuge tubes
  • Printed grid on one-way keyed lid for proper orientation and sample identification
  • Molded of impact-resistant polycarbonate with locking handle for long-term use
  • Stackable to save space
Specifications

Specifications

125 mm
151 mm
108 mm
5.32 x 4.25 x 4.94 in. (151 x 108 x 125 mm)
3 x 4
Non-filled/clear lid
Benchtop Cooler
0.2 to 0.5 mL
0°C
4.94 in.
5.32 in.
4.25 in.
3x4 compartments, non-filled, 0 deg.
<1°C, 3.5
Polycarbonate
Blue, Clear
12 Cryogenic Vials, Microcentrifuge Tubes (0.2 to 0.5 mL)
Videos
SDS
Documents

Documents

Promotions

Promotions