Thermo Scientific™ Benchtop Dewar Flasks
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with these benchtop Dewar flasks with double-walled sides, insulation and pour spouts.
$1161.48 - $2439.79
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
10-194-100A
|
Thermo Scientific™
41501000 |
1 L |
Each for $1,161.48
Case of 4 Each for $4,645.92
|
|
|||||
10-194-100B
|
Thermo Scientific™
41502000 |
2 L |
Each for $1,181.45
Case of 2 Each for $2,362.90
|
|
|||||
10-194-100C
|
Thermo Scientific™
41504000 |
4 L |
Each for $1,289.41
Case of 2 Each for $2,578.82
|
|
|||||
10-194-100D
|
Thermo Scientific™
41509000 |
10 L |
Each for $2,439.79
|
|
|||||
Description
Store samples in liquid nitrogen, dry ice-solvent mixtures or ice water for the short term with Thermo Scientific™ Benchtop Dewar Flasks. Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer. Flasks offer pour spouts and bottom grip area for easy removal of used liquids.
- High density polyethylene base and HDPE cover with polyethylene coated handle for strong, shatterproof performance
- Double-walled sides and CFC-free foam urethane insulation minimize heat leakage so samples stay cold longer
- Can be used from -196°C to +100°C
- Pour spout and bottom grip area for ease of removal of used liquids
- Vented, insulated cover allows for vapor escape but maintains sample temperature
Specifications
1 |