Learn More
Benzalkonium chloride, 50% w/w aq. soln., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B20760AP
Description
Benzalkonium chloride is a cationic surface active agent.It is used as a preservative in pharmaceutical products like eye, ear and nasal drops. It finds application in personal care products such as wet wipes, shampoos, hand sanitizers, deodorants and cosmetics. It is widely used in the chemical industry as a precipitant and emulsifier/de-emulsifier. It acts as corrosion inhibitor in the oil and gas industry pipeline by preventing the formation of sulfurous gases. It is also used in various non-consumer process like surgical disinfection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzalkonium chloride is a cationic surface active agent.It is used as a preservative in pharmaceutical products like eye, ear and nasal drops. It finds application in personal care products such as wet wipes, shampoos, hand sanitizers, deodorants and cosmetics. It is widely used in the chemical industry as a precipitant and emulsifier/de-emulsifier. It acts as corrosion inhibitor in the oil and gas industry pipeline by preventing the formation of sulfurous gases. It is also used in various non-consumer process like surgical disinfection.
Solubility
Miscible with water, methanol and acetone.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and metals.
Specifications
Benzalkonium chloride | |
0.99 | |
C21H38ClN | |
500 mL | |
4062599 | |
benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides | |
JBIROUFYLSSYDX-UHFFFAOYSA-M | |
benzyl-decyl-dimethylazanium;chloride | |
13762 |
63449-41-2 | |
None | |
MFCD00137276 | |
UN3265 | |
14,1059 | |
Miscible with water,methanol and acetone. | |
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 | |
339.99 | |
50% w/w aq. soln. |
Chemical Identifiers
63449-41-2 | |
339.99 | |
JBIROUFYLSSYDX-UHFFFAOYSA-M | |
13762 | |
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 |
C21H38ClN | |
MFCD00137276 | |
benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides | |
benzyl-decyl-dimethylazanium;chloride |
Safety and Handling
GHS H Statement
H314-H302
Causes severe skin burns and eye damage.
Harmful if swallowed.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302+H312-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 264-151-6
RTECSNumber : BO3151000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only