missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzalkonium Chloride (50% in Water), TCI America™
$42.87 - $143.54
Chemical Identifiers
| CAS | 63449-41-2 |
|---|---|
| Molecular Formula | C21H38ClN |
| Molecular Weight (g/mol) | 339.99 |
| MDL Number | MFCD00137276 |
| InChI Key | JBIROUFYLSSYDX-UHFFFAOYSA-M |
| Synonym | benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides |
| PubChem CID | 13762 |
| IUPAC Name | benzyl(dodecyl)dimethylazanium chloride |
| SMILES | [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 |
Chemical Identifiers
| 63449-41-2 | |
| 339.99 | |
| JBIROUFYLSSYDX-UHFFFAOYSA-M | |
| 13762 | |
| [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 |
| C21H38ClN | |
| MFCD00137276 | |
| benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides | |
| benzyl(dodecyl)dimethylazanium chloride |
Specifications
| 63449-41-2 | |
| C21H38ClN | |
| 25 g | |
| benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides | |
| [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1 | |
| 339.99 | |
| Liquid |
| Yellow | |
| MFCD00137276 | |
| 1760 | |
| JBIROUFYLSSYDX-UHFFFAOYSA-M | |
| benzyl(dodecyl)dimethylazanium chloride | |
| 13762 | |
| Benzalkonium Chloride (50% in Water) |
Safety and Handling
EINECSNumber : (3)-2694
RTECSNumber : BO3151000
TSCA : Yes