Learn More
Benzalkonium chloride, alkyl distribution from C8H17 to C16H33
CAS: 8001-54-5 | C9H13ClN | 170.66 g/mol
$85.14 - $2018.43
Chemical Identifiers
| CAS | 8001-54-5 |
|---|---|
| Molecular Formula | C9H13ClN |
| Molecular Weight (g/mol) | 170.66 |
| MDL Number | MFCD00145757 |
| Synonym | Alkyl-benzyl-dimethylammonium chloride |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC215411000
|
Thermo Scientific Chemicals
215411000 |
100 g | Glass bottle |
Each for $85.14
|
|
||||
|
AC215410010
|
Thermo Scientific Chemicals
215410010 |
1 kg | Glass bottle |
Each for $526.12
|
|
||||
|
AC215410051
|
Thermo Scientific Chemicals
215410051 |
5 kg | Plastic Bottle |
Each for $2,018.43
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 40.0°C | |
| Solid or Thick Gel | |
| C9H13ClN | |
| MFCD00145757 | |
| Alkyl-benzyl-dimethylammonium chloride | |
| 170.66 | |
| Authentic | |
| 0.98 | |
| Benzalkonium chloride |
| 8001-54-5 | |
| 100 g | |
| C6H5CH2N(CH3)2RClR=C8H17toC16H33 | |
| 15, 1061 | |
| Solubility in water: 4000g/L (20°C). Other solubilities: soluble in alcohol and acetone,slightly soluble in benzene,almost insoluble in ether | |
| 95.0 to 104.0% (anhydrous basis) | |
| Glass bottle | |
| 0.9800g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Very toxic to aquatic life.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Do not get in eyes,on skin,or on clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes.
GHS Signal Word: Danger
RUO – Research Use Only