missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzalkonium chloride, Thermo Scientific Chemicals

Cationic surface active agent

$53.30 - $287.84

Chemical Identifiers

CAS 63449-41-2
Molecular Formula C21H38ClN
Molecular Weight (g/mol) 339.99
MDL Number MFCD00137276
InChI Key JBIROUFYLSSYDX-UHFFFAOYSA-M
Synonym benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
PubChem CID 13762
IUPAC Name benzyl-decyl-dimethylazanium;chloride
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA4133914
View Documents
Thermo Scientific Chemicals
04133914
25 g
Each for $53.30
 
AA4133922
View Documents
Thermo Scientific Chemicals
04133922
100 g
Each for $91.41
 
AA4133936
View Documents
Thermo Scientific Chemicals
04133936
500 g
Each for $287.84
 
Description

Description

Cationic surface active agentBenzalkonium chloride is used in textile dyeing, softeners, antistatic agents, emulsifiers and conditioner. It acts as a cationic detergent. It is also used as a preservative in pharmaceutical products like eye, ear and nasal drops and sprays. Further, it is an active ingredient in personal care products such as hand sanitizers, shampoos, deodorants and wet wipes. In addition to this, it is used in a study to assess its action on epithelia conjunctival cells in vitro.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

63449-41-2
339.99
JBIROUFYLSSYDX-UHFFFAOYSA-M
13762
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
C21H38ClN
MFCD00137276
benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
benzyl-decyl-dimethylazanium;chloride

Specifications

63449-41-2
MFCD00137276
benzyldimethyldecylammonium chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride, benzenemethanaminium, n-decyl-n,n-dimethyl-, chloride 1:1, quaternary ammonium compounds, benzyl-c8-18-alkyldimethyl, chlorides, roccal, benzyl decyl dimethylazanium chloride, tret-o-lite xc 511, ccris 4586, benzyl decyl dimethylammonium chloride, c8-18-alkydimethylbenzyl ammonium chlorides
JBIROUFYLSSYDX-UHFFFAOYSA-M
benzyl-decyl-dimethylazanium;chloride
13762
Benzalkonium Chloride
C21H38ClN
25 g
Miscible with water,acetone,ether and alcohol. Slightly miscible with benzene.
[Cl-].CCCCCCCCCCCC[N+](C)(C)CC1=CC=CC=C1
339.99
Semi-solid
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302+H312-H314-H335

DOTInformation : Hazard Class: 8; Packaging Group: III

EINECSNumber : 264-151-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only