Learn More
Benzamide oxime, 97%, Thermo Scientific™
Chemical Identifiers
CAS | 613-92-3 |
---|---|
Molecular Formula | C7H8N2O |
Molecular Weight (g/mol) | 136.15 |
MDL Number | MFCD00031485 |
InChI Key | MXOQNVMDKHLYCZ-UHFFFAOYSA-N |
Synonym | benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim |
PubChem CID | 7259353 |
ChEBI | CHEBI:83354 |
IUPAC Name | N'-hydroxybenzenecarboximidamide |
SMILES | N\C(=N/O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC453280010
|
Thermo Scientific Chemicals
453280010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
AC453280050
|
Thermo Scientific Chemicals
453280050 |
5g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
613-92-3 | |
136.15 | |
MXOQNVMDKHLYCZ-UHFFFAOYSA-N | |
7259353 | |
N'-hydroxybenzenecarboximidamide |
C7H8N2O | |
MFCD00031485 | |
benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim | |
CHEBI:83354 | |
N\C(=N/O)C1=CC=CC=C1 |
Specifications
613-92-3 | |
94% min. (HPLC) | |
C7H8N2O | |
MFCD00031485 | |
benzamidoxime, benzamide oxime, n'-hydroxybenzimidamide, benzohydroxamamide, n-hydroxybenzamidine, phenylhydroxamidine, n-hydroxybenzenecarboximidamide, ccris 2953, n'-hydroxybenzamidine, benzamidoxim | |
N\C(=N/O)C1=CC=CC=C1 | |
136.15 | |
CHEBI:83354 | |
97% |
Authentic | |
Glass Bottle | |
C6H5C(NH2)=NOH | |
1g | |
MXOQNVMDKHLYCZ-UHFFFAOYSA-N | |
N'-hydroxybenzenecarboximidamide | |
7259353 | |
136.15 | |
Benzamide oxime |
Safety and Handling
GHS H Statement
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POIS
EINECSNumber : 210-361-8
RUO â Research Use Only