missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzamidine hydrochloride hydrate, 98%, water ca 10-14%, Thermo Scientific Chemicals

$50.44 - $369.67

Chemical Identifiers

CAS 206752-36-5
Molecular Formula C7H9ClN2
Molecular Weight (g/mol) 156.61
MDL Number MFCD00066285
InChI Key LZCZIHQBSCVGRD-UHFFFAOYSA-N
Synonym benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
PubChem CID 16219042
SMILES Cl.NC(=N)C1=CC=CC=C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1600206
View Documents
Thermo Scientific Chemicals
A1600206
5 g
Each for $50.44
 
AAA1600214
View Documents
Thermo Scientific Chemicals
A1600214
25 g
Each for $138.59
 
AAA1600222
View Documents
Thermo Scientific Chemicals
A1600222
100 g
Each for $369.67
 
Description

Description

Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzamidine hydrochloride hydrate is a reversible inhibitor of trypsin, trypsin-like enzymes, and serine proteases. It is a proteolytic inhibitor and apoptosis reagent.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature.
Specifications

Chemical Identifiers

206752-36-5
156.61
LZCZIHQBSCVGRD-UHFFFAOYSA-N
16219042
C7H9ClN2
MFCD00066285
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
Cl.NC(=N)C1=CC=CC=C1

Specifications

206752-36-5
C7H9ClN2
5 g
Hygroscopic
Soluble in water.
Cl.NC(=N)C1=CC=CC=C1
156.61
156.62 (Anhydrous)
Benzamidine hydrochloride hydrate
82°C to 85°C
MFCD00066285
3594959
benzamidine hydrochloride hydrate, benzenecarboximidamide hydrochloride hydrate, benzamidine hydrate hydrochloride, benzamidinium chloride, benzamidinehydrochloridehydrate, amidinobenzene hydrochloride, benzamidine hydrochloride monohydrate, acmc-20a6bw, ksc491m3d, benzamidin hydrochloride hydrate
LZCZIHQBSCVGRD-UHFFFAOYSA-N
benzenecarboximidamide;hydrate;hydrochloride
16219042
98%
water ≈10 to 14%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 216-795-4

RTECSNumber : CV6260000

TSCA : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only