Learn More
Benzanilide, 98+%, Thermo Scientific Chemicals
$68.85 - $250.19
Chemical Identifiers
CAS | 93-98-1 |
---|---|
Molecular Formula | C13H11NO |
Molecular Weight (g/mol) | 197.237 |
MDL Number | MFCD00003069 |
InChI Key | ZVSKZLHKADLHSD-UHFFFAOYSA-N |
Synonym | benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline |
PubChem CID | 7168 |
IUPAC Name | N-phenylbenzamide |
SMILES | C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1108622
|
Thermo Scientific Chemicals
A1108622 |
100 g |
N/A
|
|
||||||
AAA1108636
|
Thermo Scientific Chemicals
A1108636 |
500 g |
N/A
|
|
||||||
Description
Benzanilide is used to make dyes, active pharmaceutical ingredients and perfumes. It is also used to study the influence of beta-cyclodextrin on photorearrangement of acetanilide, benzanilide and ethyl phenyl carbonate. Further, it is used as an amide model compound to study the reaction between the amide and epoxy. In addition to this, it reacts with aryltriflates in the presence of palladium-based catalyst system to get N-(2,6-diarylbenzoyl)anilines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzanilide is used to make dyes, active pharmaceutical ingredients and perfumes. It is also used to study the influence of beta-cyclodextrin on photorearrangement of acetanilide, benzanilide and ethyl phenyl carbonate. Further, it is used as an amide model compound to study the reaction between the amide and epoxy. In addition to this, it reacts with aryltriflates in the presence of palladium-based catalyst system to get N-(2,6-diarylbenzoyl)anilines.
Solubility
Slightly soluble in diethyl ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
93-98-1 | |
197.237 | |
ZVSKZLHKADLHSD-UHFFFAOYSA-N | |
7168 | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 |
C13H11NO | |
MFCD00003069 | |
benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline | |
N-phenylbenzamide |
Specifications
93-98-1 | |
1.32 | |
Odorless | |
MFCD00003069 | |
1102980 | |
benzanilide, n-benzoylaniline, benzamide, n-phenyl, n-phenyl-benzamide, benzoic acid anilide, unii-ak1b12366o, phenyl-n-benzamide, benzanilid, benzoylanilide, n-benzoyl aniline | |
ZVSKZLHKADLHSD-UHFFFAOYSA-N | |
N-phenylbenzamide | |
7168 | |
≥98% |
162°C to 166°C | |
117°C (10 mmHg) | |
C13H11NO | |
100 g | |
14,1061 | |
Slightly soluble in diethyl ether. Insoluble in water. | |
C1=CC=C(C=C1)C(=O)NC2=CC=CC=C2 | |
197.237 | |
197.24 | |
Benzanilide |
Safety and Handling
EINECSNumber : 202-292-7
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only