Learn More
Benzeneboronic acid, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1425730
Description
Benzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzeneboronic acid is widely utilized in various cross coupling reactions for example Mizoroki-Heck and Suzuki-Miyaura coupling reactions, etc. It serves as a receptor for carbohydrates, antimicrobial agents and enzyme inhibitors in biological reactions. It acts as a reagent in palladium-catalyzed stereoselective Heck-type reaction and as a catalyst in Ni(II) pincer complex and Pd(II) pyridoxal hydrazone metallacycles. It is also employed as a sensor for neutron capture therapy for cancer, transmembrane transport and bioconjugation and labeling of proteins and cell surface.
Solubility
Slightly soluble in water.
Notes
Hygroscopic. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents. Avoid moisture exposure.
Specifications
Benzeneboronic acid | |
214°C to 219°C | |
MFCD00002103 | |
970972 | |
14,1068 | |
Slightly soluble in water. | |
OB(O)C1=CC=CC=C1 | |
121.93 | |
CHEBI:44923 | |
≥98% |
98-80-6 | |
C6H7BO2 | |
250 g | |
Hygroscopic | |
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
HXITXNWTGFUOAU-UHFFFAOYSA-N | |
phenylboronic acid | |
66827 | |
121.93 |
Chemical Identifiers
98-80-6 | |
121.93 | |
HXITXNWTGFUOAU-UHFFFAOYSA-N | |
66827 | |
phenylboronic acid |
C6H7BO2 | |
MFCD00002103 | |
benzeneboronic acid, boronic acid, phenyl, phenyl boronic acid, phenylboric acid, dihydroxyphenylborane, phenyldihydroxyborane, dihydroxy phenyl borane, phenylboronicacid, borophenylic acid, usaf bo-2 | |
CHEBI:44923 | |
OB(O)C1=CC=CC=C1 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P264-P270-P301+P312-P304+P312-P305+P351+P338-P312-P330-P501c
H302
EINECSNumber : 202-701-9
RTECSNumber : CY8575000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only