missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzenesulfonanilide 98.0+%, TCI America™
Supplier: TCI America B0029500G
Specifications
Benzenesulfonanilide | |
110°C | |
C12H11NO2S | |
500 g | |
XAUGWFWQVYXATQ-UHFFFAOYSA-N | |
N-phenylbenzenesulfonamide | |
74296 | |
≥98.0% (HPLC,N) |
1678-25-7 | |
White | |
MFCD00035919 | |
benzenesulfonanilide, benzoylsulfanilide, benzenesulfoanilide, benzensulfonanilide, benzenesulfanilide, benzenesulfonamide, n-phenyl, n-phenylbenzenesulphonamide, phenyl phenylsulfonyl amine, n-phenyl-benzenesulfonamide, chembl83148 | |
O=S(=O)(NC1=CC=CC=C1)C1=CC=CC=C1 | |
233.29 | |
233.29 | |
Crystalline Powder |
Chemical Identifiers
1678-25-7 | |
233.29 | |
XAUGWFWQVYXATQ-UHFFFAOYSA-N | |
74296 | |
O=S(=O)(NC1=CC=CC=C1)C1=CC=CC=C1 |
C12H11NO2S | |
MFCD00035919 | |
benzenesulfonanilide, benzoylsulfanilide, benzenesulfoanilide, benzensulfonanilide, benzenesulfanilide, benzenesulfonamide, n-phenyl, n-phenylbenzenesulphonamide, phenyl phenylsulfonyl amine, n-phenyl-benzenesulfonamide, chembl83148 | |
N-phenylbenzenesulfonamide |
Safety and Handling
EINECSNumber : (3)-3781
RTECSNumber : DB3500000
TSCA : No