missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzenesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 401850250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzenesulfonic acid, sodium salt | |
515-42-4 | |
100.0 | |
White | |
Authentic | |
Glass bottle | |
SO3NaC6H5 | |
25 g | |
sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate | |
MZSDGDXXBZSFTG-UHFFFAOYSA-M | |
sodium;benzenesulfonate | |
517327 | |
98% |
98% | |
97.5 | |
450.0°C | |
2% max. (105°C) | |
97.5% min. (HPLC) | |
C6H5NaO3S | |
MFCD00065179 | |
15,1071 | |
(5% in water) Clear colorless to almost colorless | |
C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+] | |
180.16 | |
180.16 | |
Crystalline Powder |
Chemical Identifiers
515-42-4 | |
180.16 | |
MZSDGDXXBZSFTG-UHFFFAOYSA-M | |
517327 | |
C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+] |
C6H5NaO3S | |
MFCD00065179 | |
sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate | |
sodium;benzenesulfonate |
Safety and Handling
HYGROSCOPIC
EINECSNumber : 208-198-2
RTECSNumber : DB7350000
TSCA : TSCA
RUO – Research Use Only