missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzenesulfonic acid, sodium salt, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 401851000

 View more versions of this product

Catalog No. AC401851000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzenesulfonic acid, sodium salt
515-42-4
100.0
White
Authentic
Glass bottle
SO3NaC6H5
100 g
sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate
MZSDGDXXBZSFTG-UHFFFAOYSA-M
sodium;benzenesulfonate
517327
98%
98%
97.5
450.0°C
2% max. (105°C)
97.5% min. (HPLC)
C6H5NaO3S
MFCD00065179
15,1071
(5% in water) Clear colorless to almost colorless
C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
180.16
180.16
Crystalline Powder

Chemical Identifiers

515-42-4
180.16
MZSDGDXXBZSFTG-UHFFFAOYSA-M
517327
C1=CC=C(C=C1)S(=O)(=O)[O-].[Na+]
C6H5NaO3S
MFCD00065179
sodium benzenesulfonate, benzenesulfonic acid sodium salt, benzenesulfonic acid, sodium salt, dispersant gn, sodium phenylsulfonate, sodium benzenesulphonate, sodium benzene sulfonate, sodium benzene monosulfate, unii-k5rm14azhx, sodium benzene monosulfonate
sodium;benzenesulfonate
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 208-198-2

RTECSNumber : DB7350000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only