Learn More
Benzenesulfonyl chloride, 99%
CAS: 98-09-9 | C6H5ClO2S | 176.62 g/mol
$143.01 - $386.02
Chemical Identifiers
| CAS | 98-09-9 |
|---|---|
| Molecular Formula | C6H5ClO2S |
| Molecular Weight (g/mol) | 176.62 |
| MDL Number | MFCD00007426 |
| InChI Key | CSKNSYBAZOQPLR-UHFFFAOYSA-N |
| Synonym | phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride |
| PubChem CID | 7369 |
| IUPAC Name | benzenesulfonyl chloride |
| SMILES | C1=CC=C(C=C1)S(=O)(=O)Cl |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC148482500
|
Thermo Scientific Chemicals
148482500 |
250 mL | Glass bottle |
Each for $143.01
|
|
||||
|
AC148480010
|
Thermo Scientific Chemicals
148480010 |
1 L | Glass bottle |
Each for $386.02
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 98-09-9 | |
| 176.62 | |
| CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
| 7369 | |
| C1=CC=C(C=C1)S(=O)(=O)Cl |
| C6H5ClO2S | |
| MFCD00007426 | |
| phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
| benzenesulfonyl chloride |
Specifications
| 98-09-9 | |
| 100.0 | |
| 1.3840g/mL | |
| 128°C | |
| 98.5% min. (GC) | |
| C6H5ClO2S | |
| C6H5SO2Cl | |
| 250 mL | |
| 01,46; 05,22; 14,27 | |
| 15,1073 | |
| Solubility in water: may decompose. Other solubilities: soluble in alcohol and ether | |
| C1=CC=C(C=C1)S(=O)(=O)Cl | |
| 176.62 | |
| 176.62 | |
| Liquid |
| 98.5 | |
| 14.5°C | |
| 251.0°C to 252.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5508 to 1.5528 | |
| MFCD00007426 | |
| 11,34 | |
| 1.384 | |
| phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
| CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
| benzenesulfonyl chloride | |
| 7369 | |
| 99% | |
| Benzenesulfonyl chloride, 99% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 202-636-6
RTECSNumber : DB8750000
TSCA : TSCA
RUO – Research Use Only