missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzenesulfonyl Chloride 99.0+%, TCI America™
$31.60 - $73.08
Chemical Identifiers
CAS | 98-09-9 |
---|---|
Molecular Formula | C6H5ClO2S |
Molecular Weight (g/mol) | 176.614 |
MDL Number | MFCD00007426 |
InChI Key | CSKNSYBAZOQPLR-UHFFFAOYSA-N |
Synonym | phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride |
PubChem CID | 7369 |
IUPAC Name | benzenesulfonyl chloride |
SMILES | C1=CC=C(C=C1)S(=O)(=O)Cl |
Chemical Identifiers
98-09-9 | |
176.614 | |
CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
7369 | |
C1=CC=C(C=C1)S(=O)(=O)Cl |
C6H5ClO2S | |
MFCD00007426 | |
phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
benzenesulfonyl chloride |
Specifications
98-09-9 | |
252°C | |
MFCD00007426 | |
2225 | |
CSKNSYBAZOQPLR-UHFFFAOYSA-N | |
benzenesulfonyl chloride | |
7369 | |
≥99.0% (GC,T) | |
Benzenesulfonyl Chloride |
Yellow | |
C6H5ClO2S | |
25 g | |
phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride | |
C1=CC=C(C=C1)S(=O)(=O)Cl | |
176.614 | |
176.61 | |
Liquid |
Safety and Handling
EINECSNumber : (3)-1893
RTECSNumber : DB8750000
TSCA : Yes