missing translation for 'onlineSavingsMsg'
Learn More

Benzil dioxime, 98%

CAS: 23873-81-6 | C14H12N2O2 | 240.26 g/mol

Supplier:  Thermo Scientific Chemicals A1068714

 View more versions of this product

Catalog No. AAA1068714

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzil dioxime
∼238°C (decomposition)
MFCD00002113
2053615
diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0
O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1
240.26
240.26
23873-81-6
C14H12N2O2
25 g
14,1079
JJZONEUCDUQVGR-VCFJNTAESA-N
N-[(Z)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
5369401
98%

Chemical Identifiers

23873-81-6
240.26
JJZONEUCDUQVGR-VCFJNTAESA-N
5369401
O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1
C14H12N2O2
MFCD00002113
diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0
N-[(Z)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 245-921-0

RTECSNumber : DD1985000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only