missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[a]pyrene, 96%, Thermo Scientific Chemicals

Extensively used in cancer research

$110.25 - $442.68

Chemical Identifiers

CAS 50-32-8
Molecular Formula C20H12
Molecular Weight (g/mol) 252.32
MDL Number MFCD00003602
InChI Key FMMWHPNWAFZXNH-UHFFFAOYSA-N
Synonym benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
PubChem CID 2336
ChEBI CHEBI:29865
IUPAC Name benzo[a]pyrene
SMILES C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA15856MC
View Documents
Thermo Scientific Chemicals
015856MC
100 mg
Each for $110.25
 
AA1585602
View Documents
Thermo Scientific Chemicals
01585602
0.5 g
Each for $442.68
 
Description

Description

Extensively used in cancer research and for GC (Gas Chromatography) and LC (Liquid Chromatography) Analysis. It is also a multipurpose intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

50-32-8
252.32
FMMWHPNWAFZXNH-UHFFFAOYSA-N
2336
benzo[a]pyrene
C20H12
MFCD00003602
benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
CHEBI:29865
C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34

Specifications

50-32-8
495°C
MFCD00003602
UN3077
benzo a pyrene, 3,4-benzopyrene, benzo pqr tetraphene, 3,4-benzpyrene, benzpyrene, benzo def chrysene, 6,7-benzopyrene, benz a pyrene, 3,4-benz a pyrene
FMMWHPNWAFZXNH-UHFFFAOYSA-N
benzo[a]pyrene
2336
252.3
Crystalline
177°C to 180°C
C20H12
100 mg
14,1103
Soluble in benzene, toluene, and xylene; Sparingly soluble in alcohol, methanol; Insoluble in water
C1=CC=C2C(=C1)C=C1C=CC3=CC=CC4=CC=C2C1=C34
252.32
CHEBI:29865
96%
Benzo[a]pyrene
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H340-H350-H360-H317
May cause genetic defects.
May cause cancer.
May damage fertility or the unborn child.
May cause an allergic skin reaction.

P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c

H317-H340-H350-H360FD

DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.

EINECSNumber : 200-028-5

RTECSNumber : DJ3675000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only