missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzo[b]furan-7-carboxylic acid, Thermo Scientific™

Chemical Identifiers

CAS 90484-22-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD10000617
InChI Key QMHILIQFOBNARN-UHFFFAOYSA-N
Synonym benzofuran-7-carboxylic acid, benzofuran-7-carboxylicacid, 7-benzofuranylcarboxylic acid, benzo b furan-7-carboxylic acid, 7-benzofurancarboxylic acid, 4ahs, 7-benzofurancarboxylicacid, akh
PubChem CID 13307983
IUPAC Name 1-benzofuran-7-carboxylic acid
SMILES OC(=O)C1=C2OC=CC2=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
CC78201DA
SDS View Documents
Thermo Scientific Chemicals
CC78201DA
1g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC78201DE
SDS View Documents
Thermo Scientific Chemicals
CC78201DE
5g Amber Glass Bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

90484-22-3
162.14
QMHILIQFOBNARN-UHFFFAOYSA-N
13307983
OC(=O)C1=C2OC=CC2=CC=C1
C9H6O3
MFCD10000617
benzofuran-7-carboxylic acid, benzofuran-7-carboxylicacid, 7-benzofuranylcarboxylic acid, benzo b furan-7-carboxylic acid, 7-benzofurancarboxylic acid, 4ahs, 7-benzofurancarboxylicacid, akh
1-benzofuran-7-carboxylic acid

Specifications

90484-22-3
C9H6O3
1g
QMHILIQFOBNARN-UHFFFAOYSA-N
1-benzofuran-7-carboxylic acid
13307983
97%
Amber Glass Bottle
MFCD10000617
benzofuran-7-carboxylic acid, benzofuran-7-carboxylicacid, 7-benzofuranylcarboxylic acid, benzo b furan-7-carboxylic acid, 7-benzofurancarboxylic acid, 4ahs, 7-benzofurancarboxylicacid, akh
OC(=O)C1=C2OC=CC2=CC=C1
162.14
162.14
Benzo[b]furan-7-carboxylic acid
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'