missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzo[b]thiophene-3-boronic Acid (contains varying amounts of Anhydride), TCI America™
$81.32 - $81.32
Chemical Identifiers
CAS | 113893-08-6 |
---|---|
Molecular Formula | C8H7BO2S |
Molecular Weight (g/mol) | 178.012 |
MDL Number | MFCD01114669 |
InChI Key | QVANIYYVZZLQJP-UHFFFAOYSA-N |
PubChem CID | 2776344 |
IUPAC Name | 1-benzothiophen-3-ylboronic acid |
SMILES | B(C1=CSC2=CC=CC=C12)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
B47741G
|
TCI America
B47741G |
1 g |
Each for $81.32
|
|
|||||
Chemical Identifiers
113893-08-6 | |
178.012 | |
QVANIYYVZZLQJP-UHFFFAOYSA-N | |
1-benzothiophen-3-ylboronic acid |
C8H7BO2S | |
MFCD01114669 | |
2776344 | |
B(C1=CSC2=CC=CC=C12)(O)O |
Specifications
113893-08-6 | |
C8H7BO2S | |
1 g | |
QVANIYYVZZLQJP-UHFFFAOYSA-N | |
1-benzothiophen-3-ylboronic acid | |
2776344 | |
Crystalline Powder |
230°C | |
MFCD01114669 | |
benzothiophene-3-boronic acid, benzo b thiophen-3-ylboronic acid, benzo b thien-3-ylboronic acid, benzo b thiophene-3-boronic acid, thianaphthene-3-boronic acid, benzothiophen-3-ylboronic acid, 1-benzothiophen-3-yl-boranediol, 3-boronobenzo b thiophene, 1-benzothien-3-ylboronic acid, 3-borono-1-benzothiophene | |
B(C1=CSC2=CC=CC=C12)(O)O | |
178.012 | |
178.01 | |
Benzo[b]thiophene-3-boronic Acid (contains varying amounts of Anhydride) |
Safety and Handling
TSCA : No