missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzo[ghi]perylene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 105550250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Benzo[ghi]perylene | |
191-24-2 | |
100.0 | |
Yellow | |
Authentic | |
Glass bottle | |
MFCD00004135 | |
benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784 | |
GYFAGKUZYNFMBN-UHFFFAOYSA-N | |
276.33 | |
CHEBI:34568 | |
98+% |
98+% | |
98.0 | |
276.0°C to 280.0°C | |
>500.0°C | |
98+% | |
C22H12 | |
25 mg | |
Solubility in water: insoluble | |
C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 | |
9117 | |
276.33 | |
Needle-like Crystals or Crystalline Powder |
Chemical Identifiers
191-24-2 | |
276.33 | |
GYFAGKUZYNFMBN-UHFFFAOYSA-N | |
9117 | |
C1=CC2=C3C(=C1)C4=CC=CC5=C4C6=C(C=C5)C=CC(=C36)C=C2 |
C22H12 | |
MFCD00004135 | |
benzo ghi perylene, 1,12-benzoperylene, 1,12-benzperylene, benzo g,h,i perylene, benzo ghi pyrilene, benzo-1,12-perylene, unii-5zme2e2q9l, ccris 784 | |
CHEBI:34568 |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
GHS Signal Word: Warning
EINECSNumber : 205-883-8
RTECSNumber : DI6200500
RUO – Research Use Only