missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing

Benzoic Acid (Crystalline/Certified ACS), Fisher Chemical
Supplier: Fisher Chemical FLA65500
Specifications
Benzoic Acid | |
121°C | |
249°C | |
≥99.5 % | |
C6H5COOH | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
C1=CC=C(C=C1)C(=O)O | |
122.123 | |
CHEBI:30746 | |
≥99.5% | |
500 g | |
Poly Bottle | |
1.3hPa at 96°C |
2.5 to 3.5 | |
65-85-0 | |
White | |
C7H6O2 | |
MFCD00002398 | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
benzoic acid | |
243 | |
122.12 | |
Certified ACS | |
0.005% max. | |
Solid |
Chemical Identifiers
65-85-0 | |
122.123 | |
WPYMKLBDIGXBTP-UHFFFAOYSA-N | |
243 | |
benzoic acid |
C7H6O2 | |
MFCD00002398 | |
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv | |
CHEBI:30746 | |
C1=CC=C(C=C1)C(=O)O |
Safety and Handling
