Learn More
Benzoic Acid, Sodium Salt, 99+%, Thermo Scientific Chemicals
$80.47 - $80.47
Chemical Identifiers
CAS | 532-32-1 |
---|---|
Molecular Formula | C7H5NaO2 |
Molecular Weight (g/mol) | 144.11 |
MDL Number | MFCD00012463 |
InChI Key | WXMKPNITSTVMEF-UHFFFAOYSA-M |
Synonym | sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746 |
PubChem CID | 517055 |
IUPAC Name | sodium;benzoate |
SMILES | [Na+].[O-]C(=O)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC148980010
|
Thermo Scientific Chemicals
148980010 |
1 kg | Plastic Bottle |
Each for $80.47
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
532-32-1 | |
144.11 | |
WXMKPNITSTVMEF-UHFFFAOYSA-M | |
517055 | |
[Na+].[O-]C(=O)C1=CC=CC=C1 |
C7H5NaO2 | |
MFCD00012463 | |
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746 | |
sodium;benzoate |
Specifications
532-32-1 | |
100.0 | |
99+% | |
C6H5CO2Na | |
09, 92 | |
15, 8718 | |
WXMKPNITSTVMEF-UHFFFAOYSA-M | |
sodium;benzoate | |
517055 | |
≥99.0% (on dried substance) | |
1 kg | |
Plastic Bottle | |
Sodium benzoate |
99.0 | |
White | |
C7H5NaO2 | |
MFCD00012463 | |
01,1044; 02,377 | |
sodium benzoate, benzoic acid, sodium salt, benzoic acid sodium salt, sobenate, antimol, benzoate sodium, benzoate of soda, benzoate, sodium, natrium benzoicum, caswell no. 746 | |
[Na+].[O-]C(=O)C1=CC=CC=C1 | |
144.11 | |
144.11 | |
Extra Pure | |
Authentic | |
Crystals, Granules, Flakes or Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RUO – Research Use Only