missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoic Acid Standard, For Elementary Analysis, MilliporeSigma™

Supplier:  Technidata 1001340005

Catalog No. M1001340005


Explore available promotions

Description

Description

Specifications

Specifications

Benzoic Acid Standard
White solid
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
C1=CC=C(C=C1)C(=O)O
122.123
CHEBI:30746
5 g
Characteristic odor
Solid
65-85-0
C7H6O2
WPYMKLBDIGXBTP-UHFFFAOYSA-N
benzoic acid
243
122.12
570°C
Glass bottle
0.001 hPa at 20°C

Chemical Identifiers

65-85-0
122.123
benzenecarboxylic acid, dracylic acid, benzeneformic acid, carboxybenzene, phenylformic acid, benzenemethanoic acid, phenylcarboxylic acid, retardex, benzoesaeure gk, benzoesaeure gv
CHEBI:30746
C1=CC=C(C=C1)C(=O)O
C7H6O2
WPYMKLBDIGXBTP-UHFFFAOYSA-N
243
benzoic acid
Videos
Safety and Handling

Safety and Handling

H302: Harmful if swallowed.
H319: Causes serious eye irritation.

SDS
Documents

Documents

Promotions

Promotions