missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 93-97-0 |
---|---|
Molecular Formula | C14H10O3 |
Molecular Weight (g/mol) | 226.23 |
MDL Number | MFCD00003073 |
InChI Key | CHIHQLCVLOXUJW-UHFFFAOYSA-N |
Synonym | benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate |
PubChem CID | 7167 |
ChEBI | CHEBI:38815 |
IUPAC Name | benzoyl benzoate |
SMILES | C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC105501000
|
Thermo Scientific Chemicals
105501000 |
100 g | Plastic bottle |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
93-97-0 | |
226.23 | |
CHIHQLCVLOXUJW-UHFFFAOYSA-N | |
7167 | |
benzoyl benzoate |
C14H10O3 | |
MFCD00003073 | |
benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate | |
CHEBI:38815 | |
C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2 |
Specifications
93-97-0 | |
100.0 | |
White | |
360.0°C | |
Authentic | |
Plastic bottle | |
(C6H5CO)2O | |
100 g | |
01,49; 05,23 | |
15,1094 | |
Solubility in water: 0,01g/L. Other solubilities: soluble in alcohol,chloroform,acetone,benzene,,ethyl acetate,toluene,xylene,ether,glacial,acetic acid,acetic anhydride,moderately soluble in petroleum ether | |
C1=CC=C(C=C1)C(=O)OC(=O)C2=CC=CC=C2 | |
226.23 | |
CHEBI:38815 | |
98% | |
Benzoic anhydride, 98% |
97.5 | |
38.0°C to 44.0°C | |
1.1900g/mL | |
>112°C | |
97.5% min. (HPLC) | |
C14H10O3 | |
MFCD00003073 | |
09,164 | |
1.19 | |
benzoic anhydride, benzoic acid, anhydride, benzoyl anhydride, benzoylbenzoate, benzoic acid anhydride, benzoic acid, 1,1'-anhydride, phenyl anhydride, benzoesaeureanhydrid, unii-9k7x34fov2, phenylcarbonyl benzoate | |
CHIHQLCVLOXUJW-UHFFFAOYSA-N | |
benzoyl benzoate | |
7167 | |
226.23 | |
Crystalline Powder or Crystals |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Danger
EINECSNumber : 202-291-1
TSCA : TSCA
RUO – Research Use Only