missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoin, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 105510025

Catalog No. AC105510025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzoin
119-53-9
100.0
1% max.
97.5% min. (GC)
C14H12O2
MFCD00004496
15, 1095
Solubility in water: sparingly soluble
OC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
212.25
CHEBI:17682
98%
98%
97.5
194.0°C (12.0 mmHg)
Authentic
Plastic bottle
C6H5CH(OH)COC6H5
2.5kg
benzoin, 2-hydroxy-2-phenylacetophenone, benzoylphenylcarbinol, benzoin tincture, +--benzoin, ethanone, 2-hydroxy-1,2-diphenyl, bitter almond oil camphor, phenylbenzoyl carbinol, 2-hydroxy-1,2-diphenylethan-1-one, alpha-hydroxybenzyl phenyl ketone
ISAOCJYIOMOJEB-UHFFFAOYNA-N
2-hydroxy-1,2-diphenylethanone
8400
212.25
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-331-3

RTECSNumber : DI1590000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only