missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoin Isobutyl Ether 94.0+%, TCI America™

Supplier:  TCI America B101525G

 View more versions of this product

Catalog No. B101525G


Explore available promotions

Description

Description

Specifications

Specifications

Benzoin Isobutyl Ether
133°C
MFCD00008933
2-Isobutoxy-2-phenylacetophenone
CC(C)COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
268.356
268.36
Liquid
22499-12-3
C18H20O2
25 g
JMVZGKVGQDHWOI-UHFFFAOYSA-N
2-(2-methylpropoxy)-1,2-diphenylethanone
90794
≥94.0% (GC)

Chemical Identifiers

22499-12-3
268.356
JMVZGKVGQDHWOI-UHFFFAOYSA-N
90794
CC(C)COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
C18H20O2
MFCD00008933
2-Isobutoxy-2-phenylacetophenone
2-(2-methylpropoxy)-1,2-diphenylethanone
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0079

RTECSNumber : KM5782265

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions