missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoin methyl ether, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1029636

 View more versions of this product

Catalog No. AAA1029636


Explore available promotions

Description

Description

Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzoin methyl ether is used in UV curing inks, wood coatings, paper coatings, optical fiber, PCB, screen printing , paper varnish and other surface coatings. It is used for anticratering in powder coatings.

Solubility
Soluble in methanol.

Notes
Store away from oxidizing agents. Store in a cool, dry condition in well sealed containers.
Specifications

Specifications

Benzoin methyl ether
46°C to 50°C
188°C to 189°C (15 mmHg)
C15H14O2
500 g
14,1093
Soluble in methanol.
COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
226.275
226.28
3524-62-7
1.128
>110°C (230°F)
MFCD00008492
2052112
benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
BQZJOQXSCSZQPS-UHFFFAOYSA-N
2-methoxy-1,2-diphenylethanone
98097
97%

Chemical Identifiers

3524-62-7
226.275
BQZJOQXSCSZQPS-UHFFFAOYSA-N
98097
COC(C1=CC=CC=C1)C(=O)C2=CC=CC=C2
C15H14O2
MFCD00008492
benzoin methyl ether, methyl benzoin, benzoin methyl ester, o-methylbenzoin, 2-methoxy-2-phenylacetophenone, nisso cure mbo, ethanone, 2-methoxy-1,2-diphenyl, acetophenone, 2-methoxy-2-phenyl, qcu 3, alpha-methoxydeoxybenzoin
2-methoxy-1,2-diphenylethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 222-538-7

RTECSNumber : AM9350000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only