missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzophenone, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 105560010

 View more versions of this product

Catalog No. AC105560010


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzophenone
98.5
47.0°C to 49.0°C
305.0°C
Authentic
Plastic bottle
(C6H5)2CO
07,411
15,11
Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2
182.22
CHEBI:41308
99%
Crystalline Powder or Flakes
119-61-9
100.0
White
143°C
98.5% min. (GC)
C13H10O
1 kg
13,108
benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
RWCCWEUUXYIKHB-UHFFFAOYSA-N
diphenylmethanone
3102
182.22
Pure

Chemical Identifiers

119-61-9
182.22
benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone
CHEBI:41308
C1=CC=C(C=C1)C(=O)C2=CC=CC=C2
C13H10O
RWCCWEUUXYIKHB-UHFFFAOYSA-N
3102
diphenylmethanone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 204-337-6

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only