Learn More
Benzophenone, 99%, pure
CAS: 119-61-9 | C13H10O | 182.22 g/mol
Supplier: Thermo Scientific Chemicals 105565000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Benzophenone | |
| 98.5 | |
| 47.0°C to 49.0°C | |
| 305.0°C | |
| Authentic | |
| Plastic bottle | |
| (C6H5)2CO | |
| 07,411 | |
| 15,11 | |
| Solubility in water: insoluble. Other solubilities: 1g/7.5mL alcohol,1g/6mL ether,soluble in chloroform,acetone and benzene | |
| C1=CC=C(C=C1)C(=O)C2=CC=CC=C2 | |
| 182.22 | |
| CHEBI:41308 | |
| 99% | |
| Crystalline Powder or Flakes |
| 119-61-9 | |
| 100.0 | |
| White | |
| 143°C | |
| 98.5% min. (GC) | |
| C13H10O | |
| 500 g | |
| 13,108 | |
| benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone | |
| RWCCWEUUXYIKHB-UHFFFAOYSA-N | |
| diphenylmethanone | |
| 3102 | |
| 182.22 | |
| Pure |
Chemical Identifiers
| 119-61-9 | |
| 182.22 | |
| benzophenone, diphenyl ketone, benzoylbenzene, phenyl ketone, methanone, diphenyl, alpha-oxoditane, ketone, diphenyl, benzene, benzoyl, alpha-oxodiphenylmethane, diphenylketone | |
| CHEBI:41308 | |
| C1=CC=C(C=C1)C(=O)C2=CC=CC=C2 |
| C13H10O | |
| RWCCWEUUXYIKHB-UHFFFAOYSA-N | |
| 3102 | |
| diphenylmethanone |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Harmful to aquatic life with long lasting effects.
GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Get medical attention/advice if you feel unwell.
Avoid release to the environment.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 204-337-6
RUO – Research Use Only