missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzophenone oxime, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 342330050

 View more versions of this product

Catalog No. AC342330050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzophenone oxime
574-66-3
100.0
98%
C13H11NO
MFCD00051461
15, 1100
Solubility in water: insoluble. Other solubilities: insoluble in hexane,soluble in benzene(warm),chloroform,,dichloromethane,ethyl acetate,methanol
C1=CC=C(C=C1)C(=NO)C2=CC=CC=C2
197.24
197.24
98%
97.5
Authentic
Glass bottle
C6H5CNOH(C6H5)
5g
benzophenone oxime, diphenylmethanone oxime, benzophenoxime, methanone, diphenyl-, oxime, diphenyl ketoxime, benzophenoneoxime, diphenylmethylene hydroxylamine, benzophenone, oxime, unii-2dlj8c37dq, diphenylketone oxime
DNYZBFWKVMKMRM-UHFFFAOYSA-N
N-benzhydrylidenehydroxylamine
11324
98%

Chemical Identifiers

574-66-3
197.24
DNYZBFWKVMKMRM-UHFFFAOYSA-N
11324
C1=CC=C(C=C1)C(=NO)C2=CC=CC=C2
C13H11NO
MFCD00051461
benzophenone oxime, diphenylmethanone oxime, benzophenoxime, methanone, diphenyl-, oxime, diphenyl ketoxime, benzophenoneoxime, diphenylmethylene hydroxylamine, benzophenone, oxime, unii-2dlj8c37dq, diphenylketone oxime
N-benzhydrylidenehydroxylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes serious eye damage.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 209-373-6

RTECSNumber : DJ1810000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only