missing translation for 'onlineSavingsMsg'
Learn More

Benzopinacol 95.0+%, TCI America™

$44.73 - $177.08

Chemical Identifiers

CAS 464-72-2
Molecular Formula C26H22O2
Molecular Weight (g/mol) 366.46
MDL Number MFCD00004448
InChI Key MFEWNFVBWPABCX-UHFFFAOYSA-N
Synonym benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
PubChem CID 94766
IUPAC Name 1,1,2,2-tetraphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B10375G
View Documents
TCI America
B10375G
5 g
Each for $44.73
Only null left
 
B103725G
View Documents
TCI America
B103725G
25 g
Each for $177.08
Only null left
 
Description

Description

Specifications

Chemical Identifiers

464-72-2
366.46
MFEWNFVBWPABCX-UHFFFAOYSA-N
94766
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
C26H22O2
MFCD00004448
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
1,1,2,2-tetraphenylethane-1,2-diol

Specifications

464-72-2
White
MFCD00004448
3077
MFEWNFVBWPABCX-UHFFFAOYSA-N
1,1,2,2-tetraphenylethane-1,2-diol
94766
≥95.0% (HPLC)
Benzopinacol
181°C
C26H22O2
5 g
benzopinacol, benzopinacole, benzopinacone, benzpinacol, benzpinacone, tetraphenylethylene glycol, tetraphenyl-1,2-ethanediol, benzophenone pinacol, 1,1,2,2-tetraphenyl-1,2-ethanediol, 1,2-ethanediol, 1,1,2,2-tetraphenyl
C1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=CC=C4)O)O
366.46
366.46
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : KI2750000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'