missing translation for 'onlineSavingsMsg'
Learn More

Benzothiazole, 97%

CAS: 95-16-9 | C7H5NS | 135.18 g/mol

$60.75 - $183.30

Chemical Identifiers

CAS 95-16-9
Molecular Formula C7H5NS
Molecular Weight (g/mol) 135.18
MDL Number MFCD00005775
InChI Key IOJUPLGTWVMSFF-UHFFFAOYSA-N
Synonym benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
PubChem CID 7222
ChEBI CHEBI:45993
IUPAC Name 1,3-benzothiazole
SMILES C1=CC=C2C(=C1)N=CS2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC146390050
SDS View Documents
Thermo Scientific Chemicals
146390050
5 mL Glass bottle
Each for $60.75
Only null left
 
AC146392500
SDS View Documents
Thermo Scientific Chemicals
146392500
250 mL Glass bottle
Each for $183.30
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

95-16-9
135.18
IOJUPLGTWVMSFF-UHFFFAOYSA-N
7222
1,3-benzothiazole
C7H5NS
MFCD00005775
benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
CHEBI:45993
C1=CC=C2C(=C1)N=CS2

Specifications

95-16-9
100.0
Brown to Brown-Yellow
231°C
Authentic
Glass bottle
1.6410 to 1.644
5 mL
1.23
benzothiazole, benzo d thiazole, benzosulfonazole, 1-thia-3-azaindene, vangard bt, benzothiazol, usaf ek-4812, benzthiazole, fema number 3256, unii-g5bw2593ep
IOJUPLGTWVMSFF-UHFFFAOYSA-N
1,3-benzothiazole
7222
135.18
Liquid
96.0
2°C
1.2300g/mL
>112°C
96% min. (GC)
C7H5NS
MFCD00005775
15,20
15, 1110
Solubility in water: slightly soluble. Other solubilities: freely soluble in alcohol and carbon disulfide
C1=CC=C2C(=C1)N=CS2
135.18
CHEBI:45993
97%
Benzothiazole
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 202-396-2

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only