Learn More
Benzotrichloride, 98%, Thermo Scientific Chemicals
$1170.99 - $1170.99
Chemical Identifiers
CAS | 98-07-7 |
---|---|
Molecular Formula | C7H5Cl3 |
Molecular Weight (g/mol) | 195.47 |
MDL Number | MFCD00000786 |
InChI Key | XEMRAKSQROQPBR-UHFFFAOYSA-N |
Synonym | benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane |
PubChem CID | 7367 |
ChEBI | CHEBI:82274 |
IUPAC Name | trichloromethylbenzene |
SMILES | ClC(Cl)(Cl)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA156520E
|
Thermo Scientific Chemicals
A156520E |
2500 g |
Each for $1,170.99
|
|
|||||
Description
It finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt finds its use in other preparations of substituted benzophenones, for benzoylation of phenols under basic, phase-transfer conditions. D,D,D-Trichlorotoluene is an intermediate, used exclusively in the industrial production of other intermediates such as Benzoylchloride, Benzotrifluoride and 2,4-Dihydroxybenzophenone. α,α,α-Trichlorotoluene is also used in determination of chlorinated toluenes in effluents of wastewater treatment plants and textile industries by closed loop stripping analysis.
Solubility
Solubility in water is 0.1 g/L at 20°C.
Notes
Special handling precautions required. View MSDS prior to purchase. MSDS are available online at www.alfa.com
Chemical Identifiers
98-07-7 | |
195.47 | |
XEMRAKSQROQPBR-UHFFFAOYSA-N | |
7367 | |
trichloromethylbenzene |
C7H5Cl3 | |
MFCD00000786 | |
benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane | |
CHEBI:82274 | |
ClC(Cl)(Cl)C1=CC=CC=C1 |
Specifications
98-07-7 | |
1.38 | |
97°C (207°F) | |
1.557 | |
2500 g | |
508152 | |
14,1109 | |
XEMRAKSQROQPBR-UHFFFAOYSA-N | |
trichloromethylbenzene | |
7367 | |
195.48 | |
Benzotrichloride |
-8°C to -0.5°C | |
90°C to 91°C (15 mmHg) | |
C7H5Cl3 | |
MFCD00000786 | |
UN2226 | |
Moisture sensitive | |
benzotrichloride, trichloromethyl benzene, alpha,alpha,alpha-trichlorotoluene, phenylchloroform, toluene trichloride, benzenyl trichloride, benzene, trichloromethyl, benzyl trichloride, benzylidyne chloride, phenyltrichloromethane | |
ClC(Cl)(Cl)C1=CC=CC=C1 | |
195.47 | |
CHEBI:82274 | |
98% |
Safety and Handling
GHS H Statement
H330-H350-H314-H318-H302-H335
Fatal if inhaled.
May cause cancer.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
P201-P202-P261-P264b-P270-P271-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P310-P311-P330-P332+P313-P362-P501c
H302-H315-H318-H331-H335-H350
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: BENZOTRICHLORIDE
EINECSNumber : 202-634-5
RTECSNumber : XT9275000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only