missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzoyl chloride, 98+%, ACS reagent, Thermo Scientific Chemicals

$70.46 - $875.48

Chemical Identifiers

CAS 98-88-4
Molecular Formula C7H5ClO
Molecular Weight (g/mol) 140.57
MDL Number MFCD00000653
InChI Key PASDCCFISLVPSO-UHFFFAOYSA-N
Synonym benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PubChem CID 7412
ChEBI CHEBI:82275
IUPAC Name benzoyl chloride
SMILES C1=CC=C(C=C1)C(=O)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC402030050
SDS View Documents
Thermo Scientific Chemicals
402030050
5 g Glass bottle N/A
 
AC402035000
SDS View Documents
Thermo Scientific Chemicals
402035000
500 g Glass bottle N/A
 
AC402030025
SDS View Documents
Thermo Scientific Chemicals
402030025
2.5 kg Glass bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

98-88-4
140.57
PASDCCFISLVPSO-UHFFFAOYSA-N
7412
benzoyl chloride
C7H5ClO
MFCD00000653
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
CHEBI:82275
C1=CC=C(C=C1)C(=O)Cl

Specifications

98-88-4
100.0
1.2100g/mL
93°C
Authentic
Glass bottle
1.5520 to 1.5540
MFCD00000653
09, 182
1.21
benzenecarbonyl chloride, benzoic acid, chloride, benzoylchloride, alpha-chlorobenzaldehyde, benzoic acid chloride, benzoylchlorid, benzaldehyde, alpha-chloro, a-chlorobenzaldehyde, ccris 802, unii-vty8706w36
PASDCCFISLVPSO-UHFFFAOYSA-N
benzoyl chloride
7412
CHEBI:82275
98+%
Liquid
98.0
-1.0°C
198.0°C
0.005% max.
98+%
C7H5ClO
C6H5COCl
5 g
01,50; 05,24; 13,219
15, 1115
Solubility in water: reacts, 2 mg/mL. Other solubilities: decomposes in alcohols and in water, soluble in ether, benzene; miscible with oils,, carbon disulfide, ccl4
C1=CC=C(C=C1)C(=O)Cl
140.57
0.0012 Pas (30°C)
140.57
ACS Reagent
Benzoyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Toxic if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 202-710-8

RTECSNumber : DM6600000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only