Learn More
Benzoyl fluoride, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1146406
Description
Benzoyl fluoride is used as a mild fluorinating agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzoyl fluoride is used as a mild fluorinating agent.
Solubility
Hydrolyses in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. It is sensitive to moisture. Store at room temperature.
Specifications
Benzoyl fluoride | |
∼-28°C | |
159°C to 161°C | |
Pungent | |
1.496 | |
5 g | |
1904977 | |
benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
HPMLGNIUXVXALD-UHFFFAOYSA-N | |
benzoyl fluoride | |
67999 | |
98% |
455-32-3 | |
1.14 | |
48°C (118°F) | |
C7H5FO | |
MFCD00000364 | |
UN2920 | |
Moisture sensitive | |
Hydrolyses in water. | |
FC(=O)C1=CC=CC=C1 | |
124.11 | |
124.12 |
Chemical Identifiers
455-32-3 | |
124.11 | |
HPMLGNIUXVXALD-UHFFFAOYSA-N | |
67999 | |
FC(=O)C1=CC=CC=C1 |
C7H5FO | |
MFCD00000364 | |
benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
benzoyl fluoride |
Safety and Handling
GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H226-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, FLAMMABLE, N.O.S.
EINECSNumber : 207-244-9
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only