missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzoyl Fluoride 98.0+%, TCI America™
Supplier: TCI America B11155G
Specifications
Benzoyl Fluoride | |
-28°C | |
C7H5FO | |
5 g | |
benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
FC(=O)C1=CC=CC=C1 | |
124.11 | |
124.11 | |
Liquid |
455-32-3 | |
160°C | |
MFCD00000364 | |
2920 | |
HPMLGNIUXVXALD-UHFFFAOYSA-N | |
benzoyl fluoride | |
67999 | |
≥98.0% (GC,T) |
Chemical Identifiers
455-32-3 | |
124.11 | |
HPMLGNIUXVXALD-UHFFFAOYSA-N | |
67999 | |
FC(=O)C1=CC=CC=C1 |
C7H5FO | |
MFCD00000364 | |
benzoylfluoride, unii-lor25i34hd, lor25i34hd, benzoylfluorid, acmc-1alcy, hpmlgniuxvxald-uhfffaoysa | |
benzoyl fluoride |
Safety and Handling
TSCA : Yes