missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Benzydamine hydrochloride, Thermo Scientific Chemicals

$75.78 - $217.32

Chemical Identifiers

CAS 132-69-4
Molecular Formula C19H24ClN3O
Molecular Weight (g/mol) 345.871
MDL Number MFCD00078957
InChI Key HNNIWKQLJSNAEQ-UHFFFAOYSA-N
Synonym benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
PubChem CID 65464
IUPAC Name 3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
SMILES CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ6118006
View Documents
Thermo Scientific Chemicals
J6118006
5 g
Each for $75.78
 
AAJ6118014
View Documents
Thermo Scientific Chemicals
J6118014
25 g
Each for $217.32
 
Description

Description

Nonsteroidal analgesic and anti-inflammatory compound. A prostaglandin synthase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Nonsteroidal analgesic and anti-inflammatory compound
  • A prostaglandin synthase inhibitor
Specifications

Chemical Identifiers

132-69-4
345.871
HNNIWKQLJSNAEQ-UHFFFAOYSA-N
65464
CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl
C19H24ClN3O
MFCD00078957
benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride

Specifications

132-69-4
White
MFCD00078957
14,1122
Soluble in water,ethanol,chloroform,n-butanol or DMSO /n
CN(C)CCCOC1=NN(C2=CC=CC=C21)CC3=CC=CC=C3.Cl
345.871
345.87
Benzydamine hydrochloride
156°C to 161°C
C19H24ClN3O
5 g
benzydamine hydrochloride, benzydamine hcl, benalgin, benzindamine hydrochloride, benzidamine hydrochloride, benzidan, benzyrin, difflam, dorinamin, enzamin
HNNIWKQLJSNAEQ-UHFFFAOYSA-N
3-(1-benzylindazol-3-yl)oxy-N,N-dimethylpropan-1-amine;hydrochloride
65464
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H319
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P363-P501c

H302+H312+H332-H319

EINECSNumber : 205-076-0

RTECSNumber : NK7875000

TSCA : No

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only